missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxyindole, TCI America™

$53.47 - $155.84

Chemical Identifiers

CAS 20289-26-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
InChI Key LJFVSIDBFJPKLD-UHFFFAOYSA-N
Synonym 4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 4-benzyloxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
PubChem CID 88465
IUPAC Name 4-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28111G View Documents TCI America
B28111G
1g Each for $53.47
 
B28115G View Documents TCI America
B28115G
5g Each for $155.84
 
Specifications

Chemical Identifiers

20289-26-3
223.275
4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 4-benzyloxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
4-phenylmethoxy-1H-indole
C15H13NO
LJFVSIDBFJPKLD-UHFFFAOYSA-N
88465
C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3

Specifications

Undesignated
1g
C15H13NO
LJFVSIDBFJPKLD-UHFFFAOYSA-N
4-phenylmethoxy-1H-indole
88465
Crystal-Powder at 20°C
4-Benzyloxyindole
62°C
20289-26-3
4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 4-benzyloxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
223.275
223.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No