missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyindole, 99%

$49.81 - $147.27

Chemical Identifiers

CAS 20289-26-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00047200
InChI Key LJFVSIDBFJPKLD-UHFFFAOYSA-N
Synonym 4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 4-benzyloxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
PubChem CID 88465
IUPAC Name 4-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL17147MD View Documents Alfa Aesar™
L17147MD
250mg Each for $49.81
 
AAL1714703 View Documents Alfa Aesar™
L1714703
1g Each for $147.27
 
Specifications

Chemical Identifiers

20289-26-3
223.275
LJFVSIDBFJPKLD-UHFFFAOYSA-N
88465
C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
C15H13NO
MFCD00047200
4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 4-benzyloxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
4-phenylmethoxy-1H-indole

Specifications

59°C to 61°C
20289-26-3
MFCD00047200
4-benzyloxyindole, 4-benzyloxy-1h-indole, 4-phenylmethoxy-1h-indole, 4-benzyloxy-1h-indole, 1h-indole, 4-phenylmethoxy, 4-benzyloxy-indole, 4-phenylmethoxy indole, 4-benzoxyindole, 4-benzyloxy-indol, 4-benzyloxyindole;
C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
223.275
223.28
4-Benzyloxyindole
250mg
C15H13NO
183150
LJFVSIDBFJPKLD-UHFFFAOYSA-N
4-phenylmethoxy-1H-indole
88465
99%