missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyindole-3-carboxaldehyde, 98%

$85.14 - $246.74

Chemical Identifiers

CAS 7042-71-9
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.285
MDL Number MFCD00152002
InChI Key WURDDLFAUSUSLT-UHFFFAOYSA-N
Synonym 4-benzyloxyindole-3-carboxaldehyde, 4-benzyloxyindole-3-carbaldehyde, 4-benzyloxy-1h-indole-3-carbaldehyde, 4-phenylmethoxy-1h-indole-3-carboxaldehyde, 4-phenylmethoxy indole-3-carbaldehyde, 4-benayloxyindole-3-carboxaldehyde, pubchem7231, acmc-1bcm9, 4-benzyloxy-3-formylindole, 4-benzoxy-1h-indole-3-carbaldehyde
PubChem CID 7015846
IUPAC Name 4-phenylmethoxy-1H-indole-3-carbaldehyde
SMILES C1=CC=C(C=C1)COC2=CC=CC3=C2C(=CN3)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1756703 View Documents Alfa Aesar™
L1756703
1g Each for $246.74
 
AAL17567MD View Documents Alfa Aesar™
L17567MD
250mg Each for $85.14
 
Specifications

Chemical Identifiers

7042-71-9
251.285
WURDDLFAUSUSLT-UHFFFAOYSA-N
7015846
C1=CC=C(C=C1)COC2=CC=CC3=C2C(=CN3)C=O
C16H13NO2
MFCD00152002
4-benzyloxyindole-3-carboxaldehyde, 4-benzyloxyindole-3-carbaldehyde, 4-benzyloxy-1h-indole-3-carbaldehyde, 4-phenylmethoxy-1h-indole-3-carboxaldehyde, 4-phenylmethoxy indole-3-carbaldehyde, 4-benayloxyindole-3-carboxaldehyde, pubchem7231, acmc-1bcm9, 4-benzyloxy-3-formylindole, 4-benzoxy-1h-indole-3-carbaldehyde
4-phenylmethoxy-1H-indole-3-carbaldehyde

Specifications

167°C
7042-71-9
MFCD00152002
4-benzyloxyindole-3-carboxaldehyde, 4-benzyloxyindole-3-carbaldehyde, 4-benzyloxy-1h-indole-3-carbaldehyde, 4-phenylmethoxy-1h-indole-3-carboxaldehyde, 4-phenylmethoxy indole-3-carbaldehyde, 4-benayloxyindole-3-carboxaldehyde, pubchem7231, acmc-1bcm9, 4-benzyloxy-3-formylindole, 4-benzoxy-1h-indole-3-carbaldehyde
C1=CC=C(C=C1)COC2=CC=CC3=C2C(=CN3)C=O
251.285
251.28
4-Benzyloxyindole-3-carboxaldehyde
1g
C16H13NO2
1537328
WURDDLFAUSUSLT-UHFFFAOYSA-N
4-phenylmethoxy-1H-indole-3-carbaldehyde
7015846
98%