Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Benzyloxycarbonylamino)cyclohexanone, 97%

$105.92 - $379.70

Chemical Identifiers

Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanone
CAS 16801-63-1
Molecular Formula C14H17NO3
Formula Weight 247.29
MDL Number MFCD06658393
Synonym 4-(Cbz-amino)cyclohexanone; N-Benzyloxycarbonyl-4-oxocyclohexylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2601503

  SDS
alfa aesar™
H2601503
1g Each for $105.92

AAH2601506

  SDS
alfa aesar™
H2601506
5g Each for $379.70
Description & Specifications

Specifications

Formula Weight 247.29
Odor Odorless
CAS 16801-63-1
Quantity 1g
Melting Point 81° to 83°C
Assay 97%
Molecular Formula C14H17NO3
Synonym 4-(Cbz-amino)cyclohexanone; N-Benzyloxycarbonyl-4-oxocyclohexylamine
MDL Number MFCD06658393
Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanone