Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Benzyloxycarbonylamino)cyclohexanol, 97%, predominantly trans

$121.59 - $438.47

Chemical Identifiers

Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanol
CAS 16801-62-0
Name Note predominantly trans
Molecular Formula C14H19NO3
Formula Weight 249.31
MDL Number MFCD02728352
Synonym 4-(Cbz-amino)cyclohexanol; N-Benzyloxycarbonyl-4-hydroxycyclohexylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2601606

  SDS
alfa aesar™
H2601606
5g Each for $121.59

AAH2601614

  SDS
alfa aesar™
H2601614
25g Each for $438.47
Description & Specifications

Specifications

Formula Weight 249.31
Name Note predominantly trans
Melting Point 162° to 164°C
Molecular Formula C14H19NO3
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanol
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Quantity 5g
Odor Odorless
MDL Number MFCD02728352
Health Hazard 1 Warning
CAS 16801-62-0
Assay 97%
Synonym 4-(Cbz-amino)cyclohexanol; N-Benzyloxycarbonyl-4-hydroxycyclohexylamine
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.