Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Benzyloxycarbonylamino)butyric acid, 98%

Chemical Identifiers

Chemical Name or Material 4-(Benzyloxycarbonylamino)butyric acid
CAS 5105-78-2
Formula Weight 237.26
MDL Number MFCD00055835
Beilstein 1884426
Synonym Z-GABA-OH; N-Cbz-4-aminobutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6685706

 
alfa aesar™
H6685706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6685714

 
alfa aesar™
H6685714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6685722

 
alfa aesar™
H6685722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.26
Synonym Z-GABA-OH; N-Cbz-4-aminobutyric acid
Chemical Name or Material 4-(Benzyloxycarbonylamino)butyric acid
CAS 5105-78-2
MDL Number MFCD00055835
Assay 98%
RTECS Number ES8137200
Physical Form Solid
Quantity 5g
Beilstein 1884426