Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Benzyloxycarbonyl)benzeneboronic acid, 95%

$89.96 - $89.96

Chemical Identifiers

Chemical Name or Material 4-(Benzyloxycarbonyl)benzeneboronic acid
CAS 184000-11-1
Molecular Formula C14H13BO4
Formula Weight 256.06
MDL Number MFCD02179442
Synonym 4-(Benzyloxycarbonyl)phenylboronic acid; Benzyl 4-boronobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27675MC

  SDS
alfa aesar™
H27675MC
100mg Each for $89.96
Description & Specifications

Specifications

Formula Weight 256.06
Assay 95%
CAS 184000-11-1
Quantity 100mg
Chemical Name or Material 4-(Benzyloxycarbonyl)benzeneboronic acid
Molecular Formula C14H13BO4
MDL Number MFCD02179442
Synonym 4-(Benzyloxycarbonyl)phenylboronic acid; Benzyl 4-boronobenzoate
Melting Point 187° to 192°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.