Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxycarbonyl-2-piperazinone, 97%

$85.89 - $281.69

Chemical Identifiers

Chemical Name or Material 4-Benzyloxycarbonyl-2-piperazinone
CAS 78818-15-2
Molecular Formula C12H14N2O3
Formula Weight 234.25
MDL Number MFCD00173924
Synonym 3-Oxopiperazine-1-carboxylic acid benzyl ester; 1-Z-3-oxopiperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2703403

 
alfa aesar™
H2703403
1g Each for $85.89

AAH2703406

 
alfa aesar™
H2703406
5g Each for $281.69
Description & Specifications

Specifications

Formula Weight 234.25
Chemical Name or Material 4-Benzyloxycarbonyl-2-piperazinone
Assay 97%
Molecular Formula C12H14N2O3
MDL Number MFCD00173924
CAS 78818-15-2
Synonym 3-Oxopiperazine-1-carboxylic acid benzyl ester; 1-Z-3-oxopiperazine
Melting Point 120° to 122°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.