missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxybenzyl Alcohol 95.0+%, TCI America™

Manufacturer:  TCI America B160225G

 View more versions of this product

Catalog No. B160225G



Specifications

Specifications

4-Benzyloxybenzyl Alcohol
86°C
836-43-1
MFCD00004654
OEBIVOHKFYSBPE-UHFFFAOYSA-N
(4-phenylmethoxyphenyl)methanol
70043
214.26
≥95.0% (GC)
White-Yellow
25g
C14H14O2
4-benzyloxy benzyl alcohol
C1=CC=C(C=C1)COC2=CC=C(C=C2)CO
214.264
CHEBI:29486
Crystal-Powder at 20°C

Chemical Identifiers

836-43-1
214.264
OEBIVOHKFYSBPE-UHFFFAOYSA-N
70043
(4-phenylmethoxyphenyl)methanol
C14H14O2
MFCD00004654
4-benzyloxy benzyl alcohol
CHEBI:29486
C1=CC=C(C=C1)COC2=CC=C(C=C2)CO
Safety and Handling

Safety and Handling

TSCA : No

Documents