Please login to your online account to display your discounted pricing

4-Benzyloxybenzyl alcohol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzyl alcohol
CAS 836-43-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14H14O2
Linear Formula C6H5CH2OC6H4CH2OH
Formula Weight 214.26
MDL Number MFCD00004654
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC181961000

  SDS
thermo fisher scientific
AC181961000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC181960100

  SDS
thermo fisher scientific
AC181960100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC181960250

  SDS
fisher scientific acros
181960250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 214.26
Quantity 100g
Color Off-white
Melting Point 84° to 87°C
CAS Max % 100.0
Chemical Name or Material 4-Benzyloxybenzyl alcohol
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2OH
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C14H14O2
MDL Number MFCD00004654
CAS 836-43-1
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.