Alfa Aesar™ 4-Benzyloxybenzyl alcohol, 98+%
Manufacturer:  Alfa Aesar™ A1494930
 View more versions of this product
Catalog No. AAA1494930
Specifications
Specifications
4-Benzyloxybenzyl alcohol
250g
C14H14O2
1877819
OEBIVOHKFYSBPE-UHFFFAOYSA-N
(4-phenylmethoxyphenyl)methanol
70043
214.26
84°C to 88°C
836-43-1
MFCD00004654
4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
C1=CC=C(C=C1)COC2=CC=C(C=C2)CO
214.264
CHEBI:29486
≥98%
Chemical Identifiers
836-43-1
214.264
OEBIVOHKFYSBPE-UHFFFAOYSA-N
70043
(4-phenylmethoxyphenyl)methanol
C14H14O2
MFCD00004654
4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
CHEBI:29486
C1=CC=C(C=C1)COC2=CC=C(C=C2)CO
Documents