Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxybenzophenone, polymer-supported, 0.8-1.1 mmol/g on Wang resin

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzophenone
Name Note polymer-supported, 0.8 to 1.1mmol/g on Wang resin
MDL Number MFCD03454175
Synonym Benzophenone Wang resin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1947203

 
alfa aesar™
L1947203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947206

 
alfa aesar™
L1947206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947209

 
alfa aesar™
L1947209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Benzyloxybenzophenone
Name Note polymer-supported, 0.8 to 1.1mmol/g on Wang resin
MDL Number MFCD03454175
Synonym Benzophenone Wang resin
Quantity 1g