Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxybenzophenone, 98+%

$252.68 - $252.68

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzophenone
CAS 54589-41-2
Molecular Formula C20H16O2
Formula Weight 288.35g/mol
MDL Number MFCD00017508
Beilstein 2464631
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0725114

 
alfa aesar™
L07251-14
25g Each for $252.68
Description & Specifications

Specifications

Melting Point 80° to 84°C
Quantity 25g
Chemical Name or Material 4-Benzyloxybenzophenone
CAS 54589-41-2
Assay ≥98%
Molecular Formula C20H16O2
Formula Weight 288.35g/mol
MDL Number MFCD00017508
Beilstein 2464631