Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxybenzonitrile, 98%

$64.18 - $216.97

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzonitrile
CAS 52805-36-4
Molecular Formula C14H11NO
Formula Weight 209.25
MDL Number MFCD00079701
Beilstein 2372698
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1925103

  SDS
alfa aesar™
L1925103
1g Each for $64.18

AAL1925106

  SDS
alfa aesar™
L1925106
5g Each for $216.97
Description & Specifications

Specifications

Formula Weight 209.25
Chemical Name or Material 4-Benzyloxybenzonitrile
Assay 98%
Health Hazard 1 Warning
CAS 52805-36-4
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Beilstein 2372698
MDL Number MFCD00079701
Molecular Formula C14H11NO
Quantity 1g
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Melting Point 95° to 98°C
Boiling Point 173° to 175°C (0.1mmHg)
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.