Learn More
4-Benzyloxybenzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1571503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Benzyloxybenzoic acid | |
187°C to 191°C | |
MFCD00016527 | |
2111640 | |
AQSCHALQLXXKKC-UHFFFAOYSA-N | |
4-phenylmethoxybenzoic acid | |
73880 | |
98% |
1486-51-7 | |
C14H12O3 | |
1 g | |
4-benzyloxybenzoic acid, 4-benzyloxy benzoic acid, benzoic acid, 4-phenylmethoxy, p-benzyloxy benzoic acid, 4-phenylmethoxy benzoic acid, 4-benzyloxy-benzoic acid, 4-benzyl-oxy benzoic acid, benzoic acid,4-phenylmethoxy, asischem a95731 | |
OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1 | |
228.25 | |
228.25 |
Chemical Identifiers
1486-51-7 | |
228.25 | |
AQSCHALQLXXKKC-UHFFFAOYSA-N | |
4-phenylmethoxybenzoic acid |
C14H12O3 | |
MFCD00016527 | |
73880 | |
OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 216-066-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only