Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxybenzeneboronic acid, 97%

$19.70 - $63.42

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzeneboronic acid
CAS 146631-00-7
Molecular Formula C13H13BO3
Formula Weight 228.06
MDL Number MFCD01075705
Synonym 4-Benzyloxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2435101

 
alfa aesar™
B2435101
0.25g Each for $19.70

AAB2435103

 
alfa aesar™
B2435103
1g Each for $63.42
Description & Specifications

Specifications

Formula Weight 228.06
Chemical Name or Material 4-Benzyloxybenzeneboronic acid
Assay 97%
Molecular Formula C13H13BO3
MDL Number MFCD01075705
CAS 146631-00-7
Synonym 4-Benzyloxyphenylboronic acid
Melting Point 193° to 198°C
Health Hazard 1 Warning
Quantity 0.25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used as electronic intermediate. It is also utilized in HPLC and NMR spectroscopy.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.