Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxybenzaldehyde, polymer-supported

$103.76 - $384.82

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzaldehyde, polymer-supported
CAS 151896-98-9
MDL Number MFCD01632880
Synonym Aldehyde Wang resin; Benzaldehyde Wang resin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1937006

 
alfa aesar™
L1937006
5g Each for $103.76

AAL1937014

 
alfa aesar™
L1937014
25g Each for $384.82
Description & Specifications

Specifications

MDL Number MFCD01632880
CAS 151896-98-9
Synonym Aldehyde Wang resin; Benzaldehyde Wang resin
Chemical Name or Material 4-Benzyloxybenzaldehyde, polymer-supported
Quantity 5g