missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Benzyloxybenzaldehyde, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 154111000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Benzyloxybenzaldehyde | |
4397-53-9 | |
100.0 | |
Authentic | |
Glass bottle | |
C6H5CH2OC6H4CHO | |
100g | |
4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin | |
ZVTWZSXLLMNMQC-UHFFFAOYSA-N | |
4-phenylmethoxybenzaldehyde | |
78109 | |
98% |
98% | |
97.5 | |
197°C to 199°C (11.0mmHg) | |
97.5% min. (GC) | |
C14H12O2 | |
MFCD00003387,MFCD01632880 | |
08, 73 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and chloroform | |
O=CC1=CC=C(OCC2=CC=CC=C2)C=C1 | |
212.25 | |
212.25 |
Chemical Identifiers
4397-53-9 | |
212.25 | |
ZVTWZSXLLMNMQC-UHFFFAOYSA-N | |
78109 | |
O=CC1=CC=C(OCC2=CC=CC=C2)C=C1 |
C14H12O2 | |
MFCD00003387,MFCD01632880 | |
4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin | |
4-phenylmethoxybenzaldehyde |
Safety and Handling
EINECSNumber : 224-527-2
TSCA : TSCA
RUO â Research Use Only