Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxybenzaldehyde, 98%

$54.69 - $717.22

Chemical Identifiers

Chemical Name or Material 4-Benzyloxybenzaldehyde
CAS 4397-53-9
Molecular Formula C14H12O2
Formula Weight 212.25
MDL Number MFCD00003387
Beilstein 1242385
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1593414

  SDS
alfa aesar™
A1593414
25g Each for $54.69

AAA1593422

  SDS
alfa aesar™
A1593422
100g Each for $159.72

AAA1593436

  SDS
alfa aesar™
A1593436
500g Each for $717.22
Description & Specifications

Specifications

Formula Weight 212.25
Chemical Name or Material 4-Benzyloxybenzaldehyde
TSCA TSCA
Assay 98%
Molecular Formula C14H12O2
MDL Number MFCD00003387
CAS 4397-53-9
Beilstein 1242385
Boiling Point 197° to 199°C (20mmHg)
Melting Point 69° to 74°C
Quantity 25g

Applications
4-Benzyloxybenzaldehyde is used in the synthesis of (5-fluoro-(2R*,3S*)-2,3-bis(4-hydroxyphenyl)pentanenitrile),an estrogen receptor β-selective ligand. It is a position isomer of the adenylyl cyclase activator 2-benzyloxybenzaldehyde.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.