Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyaniline hydrochloride, 98%

$44.01 - $133.92

Chemical Identifiers

Chemical Name or Material 4-Benzyloxyaniline hydrochloride
CAS 51388-20-6
Molecular Formula C13H13NO·HCl
Formula Weight 235.72
MDL Number MFCD00012995
Beilstein 3633307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1685614

 
alfa aesar™
A1685614
25g Each for $44.01

AAA1685622

 
alfa aesar™
A1685622
100g Each for $133.92
Description & Specifications

Specifications

Formula Weight 235.72
Chemical Name or Material 4-Benzyloxyaniline hydrochloride
Assay 98%
Molecular Formula C13H13NO·HCl
MDL Number MFCD00012995
CAS 51388-20-6
Beilstein 3633307
Melting Point ≈225°C (dec.)
Quantity 25g
RTECS Number BW7615000