missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-3-methylbenzeneboronic acid, 96%

Manufacturer:  Alfa Aesar™ H5307306

 View more versions of this product

Catalog No. AAH5307306



Specifications

Specifications

4-Benzyloxy-3-methylbenzeneboronic acid
5g
C14H15BO3
4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, 4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylphenyl boronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O
242.081
242.08
102°C to 106°C
338454-30-1
MFCD06409944
QPXFEWQLALNXEZ-UHFFFAOYSA-N
(3-methyl-4-phenylmethoxyphenyl)boronic acid
17750522
96%

Chemical Identifiers

338454-30-1
242.081
QPXFEWQLALNXEZ-UHFFFAOYSA-N
17750522
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O
C14H15BO3
MFCD06409944
4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, 4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylphenyl boronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid
(3-methyl-4-phenylmethoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents