missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-3-methoxyphenylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ L1933103

 View more versions of this product

Catalog No. AAL1933103



Specifications

Specifications

4-Benzyloxy-3-methoxyphenylacetonitrile
68°C to 70°C
1700-29-4
MFCD00016394
2702492
KSOYPRFHKIOHMY-UHFFFAOYSA-N
2-(3-methoxy-4-phenylmethoxyphenyl)acetonitrile
297963
97%
195°C (1mmHg)
1g
C16H15NO2
UN3439
4-benzyloxy-3-methoxyphenylacetonitrile, 2-4-benzyloxy-3-methoxyphenyl acetonitrile, 2-4-benzyloxy-3-methoxyphenyl acetonitrile, 4-benzyloxy-3-methoxybenzyl cyanide, 4-benzyloxy-3-methoxyphenyl acetonitrile, 2-3-methoxy-4-phenylmethoxyphenyl acetonitrile, benzeneacetonitrile, 3-methoxy-4-phenylmethoxy, acmc-209vuk, 4-benzyl-oxy-3-methoxyphenylacetonitrile, 3-methoxy-4-benzyloxyphenyl acetonitrile
COC1=C(C=CC(=C1)CC#N)OCC2=CC=CC=C2
253.301
253.3

Chemical Identifiers

1700-29-4
253.301
KSOYPRFHKIOHMY-UHFFFAOYSA-N
297963
COC1=C(C=CC(=C1)CC#N)OCC2=CC=CC=C2
C16H15NO2
MFCD00016394
4-benzyloxy-3-methoxyphenylacetonitrile, 2-4-benzyloxy-3-methoxyphenyl acetonitrile, 2-4-benzyloxy-3-methoxyphenyl acetonitrile, 4-benzyloxy-3-methoxybenzyl cyanide, 4-benzyloxy-3-methoxyphenyl acetonitrile, 2-3-methoxy-4-phenylmethoxyphenyl acetonitrile, benzeneacetonitrile, 3-methoxy-4-phenylmethoxy, acmc-209vuk, 4-benzyl-oxy-3-methoxyphenylacetonitrile, 3-methoxy-4-benzyloxyphenyl acetonitrile
2-(3-methoxy-4-phenylmethoxyphenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents