Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-3-methoxybenzyl alcohol, 98%

$104.72 - $104.72

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-3-methoxybenzyl alcohol
CAS 33693-48-0
Molecular Formula C15H16O3
Formula Weight 244.3
MDL Number MFCD00004640
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2744306

  SDS
alfa aesar™
H2744306
5g Each for $104.72
Description & Specifications

Specifications

Formula Weight 244.3
Quantity 5g
CAS 33693-48-0
Melting Point 71° to 73°C
MDL Number MFCD00004640
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C15H16O3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 1 Warning
Chemical Name or Material 4-Benzyloxy-3-methoxybenzyl alcohol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.