missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-3-methoxybenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1471622

 View more versions of this product

Catalog No. AAA1471622



Description

Description

Applications
4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N′-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

Solubility
Soluble in chloroform, methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong bases.
Specifications

Specifications

4-Benzyloxy-3-methoxybenzaldehyde
61°C to 65°C
2426-87-1
MFCD00003365
4-benzyloxy-3-methoxybenzaldehyde, 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde
COC1=C(C=CC(=C1)C=O)OCC2=CC=CC=C2
242.274
242.27
213°C to 215°C (3.5mmHg)
100g
C15H14O3
1464258
JSHLOPGSDZTEGQ-UHFFFAOYSA-N
3-methoxy-4-phenylmethoxybenzaldehyde
75506
98%

Chemical Identifiers

2426-87-1
242.274
JSHLOPGSDZTEGQ-UHFFFAOYSA-N
75506
COC1=C(C=CC(=C1)C=O)OCC2=CC=CC=C2
C15H14O3
MFCD00003365
4-benzyloxy-3-methoxybenzaldehyde, 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde
3-methoxy-4-phenylmethoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS missing translation for 'documents'