Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-3-methoxy-beta-nitrostyrene, 99%

$55.89 - $596.68

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-3-methoxy-beta-nitrostyrene
CAS 63909-38-6
Molecular Formula C16H15NO4
Formula Weight 285.31
MDL Number MFCD00031043
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2564503

  SDS
alfa aesar™
B2564503
1g Each for $55.89

AAB2564506

  SDS
alfa aesar™
B2564506
5g Each for $161.23

AAB2564514

  SDS
alfa aesar™
B2564514
25g Each for $596.68
Description & Specifications

Specifications

Formula Weight 285.31
Health Hazard 1 Warning
Assay 99%
MDL Number MFCD00031043
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 63909-38-6
Quantity 1g
Melting Point 122° to 124°C
Chemical Name or Material 4-Benzyloxy-3-methoxy-beta-nitrostyrene
Molecular Formula C16H15NO4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.