Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-3-fluorobenzeneboronic acid, 98%

$56.04 - $139.76

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-3-fluorobenzeneboronic acid
CAS 133057-83-7
Molecular Formula C13H12BFO3
Formula Weight 246.04
MDL Number MFCD00994627
Synonym 4-Benzyloxy-3-fluorophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1852003

 
alfa aesar™
L1852003
1g Each for $139.76

AAL18520MD

 
alfa aesar™
L18520MD
250mg Each for $56.04
Description & Specifications

Specifications

Formula Weight 246.04
Chemical Name or Material 4-Benzyloxy-3-fluorobenzeneboronic acid
Assay 98%
Molecular Formula C13H12BFO3
MDL Number MFCD00994627
CAS 133057-83-7
Synonym 4-Benzyloxy-3-fluorophenylboronic acid
Melting Point 129° to 131°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.