Please login to your online account to display your discounted pricing

4-Benzyloxy-3,5-dimethylbenzaldehyde, ≥98%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  346050010

 View more versions of this product

Catalog No. AC346050010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Viscous Liquid After Melting
Formula Weight 240.3
Quantity 1g
Color Clear yellow
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 4-Benzyloxy-3, 5-dimethylbenzaldehyde
Infrared Spectrum Authentic
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C16H16O2
Linear Formula C6H5CHOC6H(CH3)CH(O)
MDL Number MFCD00800683
CAS 144896-51-5
Health Hazard 1 AIR SENSITIVE

Specifications

Chemical Name or Material 4-Benzyloxy-3, 5-dimethylbenzaldehyde
Name Note 98+%
CAS 144896-51-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C16H16O2
Linear Formula C6H5CHOC6H(CH3)CH(O)
Formula Weight 240.3
MDL Number MFCD00800683
Accessories

AIR SENSITIVE

SDS & Safety Documents