Please login to your online account to display your discounted pricing

4-Benzyloxy-3,5-dimethylbenzaldehyde, ≥98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-3, 5-dimethylbenzaldehyde
Name Note 98+%
CAS 144896-51-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C16H16O2
Linear Formula C6H5CHOC6H(CH3)CH(O)
Formula Weight 240.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346050010

  SDS
thermo fisher scientific
346050010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 240.3
Physical Form Viscous Liquid After Melting
Quantity 1g
Color Clear yellow
CAS Min % 98.0
CAS Max % 100.0
Linear Formula C6H5CHOC6H(CH3)CH(O)
Assay Percent Range 98+%
Chemical Name or Material 4-Benzyloxy-3, 5-dimethylbenzaldehyde
Name Note 98+%
MDL Number MFCD00800683
Molecular Formula C16H16O2
CAS 144896-51-5
Infrared Spectrum Authentic
Health Hazard 1 AIR SENSITIVE
Safety and Handling

AIR SENSITIVE