Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-2-methoxybenzaldehyde, 98+%

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-2-methoxybenzaldehyde
CAS 58026-14-5
Molecular Formula C15H14O3
Formula Weight 242.27g/mol
MDL Number MFCD01075699
Beilstein 1880700
Synonym 4-Benzyloxy-o-anisaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1551603

 
alfa aesar™
L15516-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point ≈180°C (2mmHg)
Melting Point 93° to 98°C
Quantity 1g
Chemical Name or Material 4-Benzyloxy-2-methoxybenzaldehyde
CAS 58026-14-5
Assay ≥98%
Molecular Formula C15H14O3
Formula Weight 242.27g/mol
MDL Number MFCD01075699
Beilstein 1880700
Synonym 4-Benzyloxy-o-anisaldehyde