missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

$36.67 - $110.00

Chemical Identifiers

CAS 166744-78-1
Molecular Formula C13H12BFO3
Molecular Weight (g/mol) 246.044
MDL Number MFCD02093064
InChI Key PQWKGFALWCOVTC-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0
PubChem CID 2737786
IUPAC Name (2-fluoro-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B46381G View Documents TCI America
B46381G
1g Each for $110.00
 
B4638200MG View Documents TCI America
B4638200MG
200mg Each for $36.67
 
Specifications

Chemical Identifiers

166744-78-1
246.044
PQWKGFALWCOVTC-UHFFFAOYSA-N
2737786
B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O
C13H12BFO3
MFCD02093064
4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0
(2-fluoro-4-phenylmethoxyphenyl)boronic acid

Specifications

White
1g
C13H12BFO3
4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0
B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O
246.044
246.04
4-Benzyloxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
155°C
166744-78-1
MFCD02093064
PQWKGFALWCOVTC-UHFFFAOYSA-N
(2-fluoro-4-phenylmethoxyphenyl)boronic acid
2737786
Crystal-Powder at 20°C
Safety and Handling

Safety and Handling

TSCA : No