Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-2-fluorobenzeneboronic acid, 96%

$91.46 - $922.14

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-2-fluorobenzeneboronic acid
CAS 166744-78-1
Molecular Formula C13H12BFO3
Formula Weight 246.04
MDL Number MFCD02093064
Synonym 4-Benzyloxy-2-fluorophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1852103

  SDS
alfa aesar™
L1852103
1g Each for $262.18

AAL1852106

  SDS
alfa aesar™
L1852106
5g Each for $922.14

AAL18521MD

  SDS
alfa aesar™
L18521MD
250mg Each for $91.46
Description & Specifications

Specifications

Formula Weight 246.04
Chemical Name or Material 4-Benzyloxy-2-fluorobenzeneboronic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H12BFO3
Assay 96%
CAS 166744-78-1
Synonym 4-Benzyloxy-2-fluorophenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD02093064
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.