Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester, 95%

$384.23 - $887.49

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester
CAS 1073354-22-9
Molecular Formula C17H20BClN2O3
Formula Weight 346.62
MDL Number MFCD06798267
Synonym 4-Benzyloxy-2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2729203

  SDS
alfa aesar™
H2729203
1g Each for $887.49

AAH27292MD

  SDS
alfa aesar™
H27292MD
250mg Each for $384.23
Description & Specifications

Specifications

Formula Weight 346.62
Chemical Name or Material 4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester
Assay 95%
Molecular Formula C17H20BClN2O3
MDL Number MFCD06798267
CAS 1073354-22-9
Synonym 4-Benzyloxy-2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Melting Point 99° to 101°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.