Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-2-chloro-5-hydroxypyrimidine, 95%

$161.23 - $438.47

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-2-chloro-5-hydroxypyrimidine
CAS 885952-28-3
Molecular Formula C11H9ClN2O2
Formula Weight 236.66
MDL Number MFCD06798283
Synonym 4-Benzyloxy-2-chloro-5-pyrimidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699303

  SDS
alfa aesar™
H2699303
1g Each for $438.47

AAH26993MD

  SDS
alfa aesar™
H26993MD
250mg Each for $161.23
Description & Specifications

Specifications

Formula Weight 236.66
Molecular Formula C11H9ClN2O2
Assay 95%
MDL Number MFCD06798283
CAS 885952-28-3
Quantity 1g
Synonym 4-Benzyloxy-2-chloro-5-pyrimidinol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 4-Benzyloxy-2-chloro-5-hydroxypyrimidine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.