Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0-1.5 mmol/g on Merrifield resin

$73.08 - $244.09

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-2,6-dimethoxybenzaldehyde
Name Note polymer-supported, 1.0 to 1.5mmol/g on Merrifield resin
MDL Number MFCD02683435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1975403

  SDS
alfa aesar™
L1975403
1g Each for $73.08

AAL1975406

  SDS
alfa aesar™
L1975406
5g Each for $244.09

AAL1975414

  SDS
alfa aesar™
L1975414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD02683435
Name Note polymer-supported, 1.0 to 1.5mmol/g on Merrifield resin
Quantity 1g
Chemical Name or Material 4-Benzyloxy-2,6-dimethoxybenzaldehyde