Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxy-2,3-difluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Benzyloxy-2,3-difluorobenzeneboronic acid pinacol ester
Formula Weight 346.18
Synonym 4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6233303

 
alfa aesar™
H6233303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62333MD

 
alfa aesar™
H62333MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 346.18
Chemical Name or Material 4-Benzyloxy-2,3-difluorobenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Quantity 1g
Synonym 4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester