missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxy-2(1H)-pyridone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438090010

 View more versions of this product

Catalog No. AC438090010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Benzyloxy-2(1H)-pyridone
Glass Bottle
199.7°C to 201.7°C
53937-02-3
100.0
C12H11NO2
DOVNUEPFPBWTSV-UHFFFAOYSA-N
4-phenylmethoxy-1H-pyridin-2-one
3607724
Powder
Authentic
Brown to Yellow
1g
97.5
97.5% min. (HPLC)
2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-1,2-dihydropyridin-2-one, 4-benzyloxy-1h-pyridin-2-one, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxypyridin-2-one
C1=CC=C(C=C1)COC2=CC(=O)NC=C2
201.23
201.23
98%

Chemical Identifiers

53937-02-3
201.23
2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-1,2-dihydropyridin-2-one, 4-benzyloxy-1h-pyridin-2-one, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxypyridin-2-one
4-phenylmethoxy-1H-pyridin-2-one
C12H11NO2
DOVNUEPFPBWTSV-UHFFFAOYSA-N
3607724
C1=CC=C(C=C1)COC2=CC(=O)NC=C2
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS Documents