missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylmorpholine-2-carboxylic acid hydrochloride, 97%

$28.07 - $326.93

Chemical Identifiers

CAS 135072-15-0
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.714
MDL Number MFCD02682031
InChI Key CEDXMALCJZSQHA-UHFFFAOYSA-N
Synonym 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
PubChem CID 2776352
IUPAC Name 4-benzylmorpholine-2-carboxylic acid;hydrochloride
SMILES C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64918MD View Documents Alfa Aesar™
H64918MD
250mg Each for $28.07
 
AAH6491803 View Documents Alfa Aesar™
H6491803
1g Each for $82.04
 
AAH6491806 View Documents Alfa Aesar™
H6491806
5g Each for $326.93
 
Specifications

Chemical Identifiers

135072-15-0
257.714
CEDXMALCJZSQHA-UHFFFAOYSA-N
2776352
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl
C12H16ClNO3
MFCD02682031
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
4-benzylmorpholine-2-carboxylic acid;hydrochloride

Specifications

244°C to 245°C
135072-15-0
MFCD02682031
14403086
CEDXMALCJZSQHA-UHFFFAOYSA-N
4-benzylmorpholine-2-carboxylic acid;hydrochloride
2776352
97%
250mg
C12H16ClNO3
UN2811
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl
257.714
257.71
4-Benzylmorpholine-2-carboxylic acid hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.