Learn More
4-Benzylmorpholine-2-carboxylic acid hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6491803
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Benzylmorpholine-2-carboxylic acid hydrochloride | |
244°C to 245°C | |
MFCD02682031 | |
UN2811 | |
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l | |
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl | |
257.714 | |
257.71 |
135072-15-0 | |
C12H16ClNO3 | |
1 g | |
14403086 | |
CEDXMALCJZSQHA-UHFFFAOYSA-N | |
4-benzylmorpholine-2-carboxylic acid;hydrochloride | |
2776352 | |
97% |
Chemical Identifiers
135072-15-0 | |
257.714 | |
CEDXMALCJZSQHA-UHFFFAOYSA-N | |
2776352 | |
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl |
C12H16ClNO3 | |
MFCD02682031 | |
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l | |
4-benzylmorpholine-2-carboxylic acid;hydrochloride |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures