Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylbiphenyl, 98+%

$65.73 - $989.76

Chemical Identifiers

Chemical Name or Material 4-Benzylbiphenyl
CAS 613-42-3
Molecular Formula C19H16
Formula Weight 244.34
MDL Number MFCD00041831
Beilstein 2047066
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1539206

  SDS
alfa aesar™
A1539206
5g Each for $65.73

AAA1539214

  SDS
alfa aesar™
A1539214
25g Each for $295.54

AAA1539222

  SDS
alfa aesar™
A1539222
100g Each for $989.76
Description & Specifications

Specifications

Formula Weight 244.34
Chemical Name or Material 4-Benzylbiphenyl
Melting Point 84° to 87°C
Assay ≥98%
Boiling Point 285° to 286°C (110mmHg)
Quantity 5g
Flash Point 208°C (406°F)
Molecular Formula C19H16
MDL Number MFCD00041831
CAS 613-42-3
Beilstein 2047066