missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylbiphenyl, 98+%

Manufacturer:  Alfa Aesar™ A1539206

 View more versions of this product

Catalog No. AAA1539206



Specifications

Specifications

4-Benzylbiphenyl
285°C to 286°C (110mmHg)
5g
C19H16
2047066
AGPLQTQFIZBOLI-UHFFFAOYSA-N
1-benzyl-4-phenylbenzene
69174
≥98%
208°C (406°F)
84°C to 87°C
613-42-3
MFCD00041831
4-benzylbiphenyl, 4-phenylmethyl-1,1'-biphenyl, 1,1'-biphenyl, 4-phenylmethyl, benzylbiphenyl, p-benzylbiphenyl, 4-phenyldiphenylmethane, 1-phenyl-4-benzylbenzene, 1, 4-phenylmethyl, acmc-1bh4u, 1-benzyl-4-phenyl-benzene
C1=CC=C(C=C1)CC2=CC=C(C=C2)C3=CC=CC=C3
244.337
244.34

Chemical Identifiers

613-42-3
244.337
AGPLQTQFIZBOLI-UHFFFAOYSA-N
69174
C1=CC=C(C=C1)CC2=CC=C(C=C2)C3=CC=CC=C3
C19H16
MFCD00041831
4-benzylbiphenyl, 4-phenylmethyl-1,1'-biphenyl, 1,1'-biphenyl, 4-phenylmethyl, benzylbiphenyl, p-benzylbiphenyl, 4-phenyldiphenylmethane, 1-phenyl-4-benzylbenzene, 1, 4-phenylmethyl, acmc-1bh4u, 1-benzyl-4-phenyl-benzene
1-benzyl-4-phenylbenzene
Documents