Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylbenzeneboronic acid pinacol ester, 97%

$90.26 - $90.26

Chemical Identifiers

Chemical Name or Material 4-Benzylbenzeneboronic acid pinacol ester
CAS 911708-01-5
Molecular Formula C19H23BO2
Formula Weight 294.2
MDL Number MFCD09266180
Synonym 4-Benzylphenylboronic acid pinacol ester; 2-(4-Benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2815603

  SDS
alfa aesar™
H2815603
1g Each for $90.26
Description & Specifications

Specifications

Formula Weight 294.2
Chemical Name or Material 4-Benzylbenzeneboronic acid pinacol ester
CAS 911708-01-5
Synonym 4-Benzylphenylboronic acid pinacol ester; 2-(4-Benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Assay 97%
Molecular Formula C19H23BO2
MDL Number MFCD09266180
Melting Point 90° to 93°C
Quantity 1g