Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Benzylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

Chemical Identifiers

Chemical Name or Material 4-(Benzylaminocarbonyl)benzeneboronic acid pinacol ester
Molecular Formula C20H24BNO3
Formula Weight 337.22
MDL Number MFCD09266184
Synonym 4-(Benzylaminocarbonyl)phenylboronic acid pinacol ester; N-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2927103

 
alfa aesar™
H2927103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2927106

 
alfa aesar™
H2927106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 337.22
Chemical Name or Material 4-(Benzylaminocarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C20H24BNO3
MDL Number MFCD09266184
Synonym 4-(Benzylaminocarbonyl)phenylboronic acid pinacol ester; N-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Melting Point 109° to 113°C
Quantity 1g