missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzylamino-1-butanol, TCI America™

$73.33 - $201.67

Chemical Identifiers

CAS 59578-63-1
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.263
MDL Number MFCD00191609
InChI Key SNRCGOVPZJPRQZ-UHFFFAOYSA-N
Synonym 4-benzylamino-1-butanol, 4-benzylamino butan-1-ol, 4-benzylamino-butan-1-ol, 4-benzylamino butan-1-ol, 4-benzylamino-1-butanol, acmc-1ay6n, 4-benzylamino ;butan-1-ol, n-4-hydroxybutyl benzylamine, 1-butanol,4-phenylmethyl amino, 1-butanol, 4-phenylmethyl amino
PubChem CID 1790018
IUPAC Name 4-(benzylamino)butan-1-ol
SMILES C1=CC=C(C=C1)CNCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B159925G View Documents TCI America
B159925G
25g Each for $201.67
 
B15995G View Documents TCI America
B15995G
5g Each for $73.33
 
Specifications

Chemical Identifiers

59578-63-1
179.263
SNRCGOVPZJPRQZ-UHFFFAOYSA-N
1790018
C1=CC=C(C=C1)CNCCCCO
C11H17NO
MFCD00191609
4-benzylamino-1-butanol, 4-benzylamino butan-1-ol, 4-benzylamino-butan-1-ol, 4-benzylamino butan-1-ol, 4-benzylamino-1-butanol, acmc-1ay6n, 4-benzylamino ;butan-1-ol, n-4-hydroxybutyl benzylamine, 1-butanol,4-phenylmethyl amino, 1-butanol, 4-phenylmethyl amino
4-(benzylamino)butan-1-ol

Specifications

140°C
25g
C11H17NO
4-benzylamino-1-butanol, 4-benzylamino butan-1-ol, 4-benzylamino-butan-1-ol, 4-benzylamino butan-1-ol, 4-benzylamino-1-butanol, acmc-1ay6n, 4-benzylamino ;butan-1-ol, n-4-hydroxybutyl benzylamine, 1-butanol,4-phenylmethyl amino, 1-butanol, 4-phenylmethyl amino
C1=CC=C(C=C1)CNCCCCO
179.263
179.26
≥96.0% (GC)
Yellow
59578-63-1
MFCD00191609
SNRCGOVPZJPRQZ-UHFFFAOYSA-N
4-(benzylamino)butan-1-ol
1790018
Clear Liquid at 20°C
4-Benzylamino-1-butanol
Safety and Handling

Safety and Handling

TSCA : No