Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylamino-1-butanol, 98+%

$329.32 - $329.32

Chemical Identifiers

Chemical Name or Material 4-Benzylamino-1-butanol
CAS 59578-63-1
Molecular Formula C11H17NO
Linear Formula C6H5CH2NH(CH2)4OH
Formula Weight 179.26g/mol
MDL Number MFCD00191609
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3082109

 
alfa aesar™
H30821-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3082118

 
alfa aesar™
H30821-18
50g Each for $329.32
Description & Specifications

Specifications

Boiling Point 140°C (0.8mmHg)
Quantity 10g
Chemical Name or Material 4-Benzylamino-1-butanol
CAS 59578-63-1
Assay ≥98%
Molecular Formula C11H17NO
Linear Formula C6H5CH2NH(CH2)4OH
Formula Weight 179.26g/mol
MDL Number MFCD00191609
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.