Learn More
4-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1566406
Description
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
4-Benzyl-4-hydroxypiperidine | |
∼80°C | |
MFCD00023147 | |
1451803 | |
4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol # | |
KJZBZOFESQSBCV-UHFFFAOYSA-N | |
4-benzylpiperidin-4-ol | |
98152 | |
97% |
51135-96-7 | |
C12H17NO | |
5 g | |
Hygroscopic | |
Slightly soluble in water. | |
C1CNCCC1(CC2=CC=CC=C2)O | |
191.274 | |
191.27 |
Chemical Identifiers
51135-96-7 | |
191.274 | |
KJZBZOFESQSBCV-UHFFFAOYSA-N | |
98152 | |
C1CNCCC1(CC2=CC=CC=C2)O |
C12H17NO | |
MFCD00023147 | |
4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol # | |
4-benzylpiperidin-4-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 257-003-7
RTECSNumber : TN6836000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only