Please login to your online account to display your discounted pricing

4-Benzyl-4-hydroxypiperidine 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  105990010

 View more versions of this product

Catalog No. AC105990010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Color Yellow to brownish
Physical Form Crystalline Powder
Formula Weight 191.27
Quantity 1g
EINECS Number 257-003-7
RTECS Number TN6836000
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C12H17NO
MDL Number MFCD00023147
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material 4-Benzyl-4-hydroxypiperidine
Name Note 95%
Assay Percent Range 95%
CAS 51135-96-7
Synonym 4-Benzyl-4-piperidinol
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Benzyl-4-hydroxypiperidine
Name Note 95%
CAS 51135-96-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00023147
Synonym 4-Benzyl-4-piperidinol
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents