missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyl-4-hydroxypiperidine, 95%, Thermo Scientific™

$221.63 - $221.63

Chemical Identifiers

CAS 51135-96-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00023147
InChI Key KJZBZOFESQSBCV-UHFFFAOYSA-N
Synonym 4-benzyl-4-hydroxypiperidine, 4-benzyl-4-piperidinol, 4-benzyl-4-piperidinol #, 4-benzyl-piperidin-4-ol, 4-piperidinol, 4-benzyl, 4-piperidinol, 4-phenylmethyl, 4benzyl-piperidin-4-ol, acmc-20amoy, maybridge3_002978, pubchem16262
PubChem CID 98152
IUPAC Name 4-benzylpiperidin-4-ol
SMILES C1CNCCC1(CC2=CC=CC=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105990050
SDS View Documents Promotion Details
Thermo Fisher Scientific
105990050
5g Glass bottle
Each for $221.63
 
Description

Description

Specifications

Chemical Identifiers

51135-96-7
191.27
KJZBZOFESQSBCV-UHFFFAOYSA-N
98152
C1CNCCC1(CC2=CC=CC=C2)O
C12H17NO
MFCD00023147
4-benzyl-4-hydroxypiperidine, 4-benzyl-4-piperidinol, 4-benzyl-4-piperidinol #, 4-benzyl-piperidin-4-ol, 4-piperidinol, 4-benzyl, 4-piperidinol, 4-phenylmethyl, 4benzyl-piperidin-4-ol, acmc-20amoy, maybridge3_002978, pubchem16262
4-benzylpiperidin-4-ol

Specifications

51135-96-7
100.0
80.0°C to 85.0°C
C12H17NO
MFCD00023147
5g
KJZBZOFESQSBCV-UHFFFAOYSA-N
4-benzylpiperidin-4-ol
98152
Crystalline Powder
4-Benzyl-4-hydroxypiperidine, 95%
94.0
Brown to Yellow
94% min. (GC)
Authentic
Glass bottle
4-benzyl-4-hydroxypiperidine, 4-benzyl-4-piperidinol, 4-benzyl-4-piperidinol #, 4-benzyl-piperidin-4-ol, 4-piperidinol, 4-benzyl, 4-piperidinol, 4-phenylmethyl, 4benzyl-piperidin-4-ol, acmc-20amoy, maybridge3_002978, pubchem16262
C1CNCCC1(CC2=CC=CC=C2)O
191.27
191.27
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 257-003-7

RTECSNumber : TN6836000

SDS
Product Certifications

RUO – Research Use Only