missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyl-4-hydroxypiperidine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 105990050

 View more versions of this product

Catalog No. AC105990050



Description

Description

Specifications

Specifications

4-Benzyl-4-hydroxypiperidine
Glass bottle
80.0°C to 85.0°C
95%
94.0
94% min. (GC)
MFCD00023147
KJZBZOFESQSBCV-UHFFFAOYSA-N
4-benzylpiperidin-4-ol
98152
Crystalline Powder
Authentic
Brown to Yellow
5g
51135-96-7
100.0
C12H17NO
4-benzyl-4-hydroxypiperidine, 4-benzyl-4-piperidinol, 4-benzyl-4-piperidinol #, 4-benzyl-piperidin-4-ol, 4-piperidinol, 4-benzyl, 4-piperidinol, 4-phenylmethyl, 4benzyl-piperidin-4-ol, acmc-20amoy, maybridge3_002978, pubchem16262
C1CNCCC1(CC2=CC=CC=C2)O
191.27
191.27
95%

Chemical Identifiers

51135-96-7
191.27
KJZBZOFESQSBCV-UHFFFAOYSA-N
98152
C1CNCCC1(CC2=CC=CC=C2)O
C12H17NO
MFCD00023147
4-benzyl-4-hydroxypiperidine, 4-benzyl-4-piperidinol, 4-benzyl-4-piperidinol #, 4-benzyl-piperidin-4-ol, 4-piperidinol, 4-benzyl, 4-piperidinol, 4-phenylmethyl, 4benzyl-piperidin-4-ol, acmc-20amoy, maybridge3_002978, pubchem16262
4-benzylpiperidin-4-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 257-003-7

RTECSNumber : TN6836000

SDS Documents