missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyl-3-thiosemicarbazide, 98+%

Manufacturer:  Alfa Aesar™ L1092006

 View more versions of this product

Catalog No. AAL1092006



Description

Description

Applications
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

4-Benzyl-3-thiosemicarbazide
5g
C8H11N3S
UN2811
4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea
C1=CC=C(C=C1)CNC(=S)NN
181.257
181.26
128°C to 131°C
13431-41-9
MFCD00025145
2096431
ZTRUHAVBRPABTK-UHFFFAOYSA-N
1-amino-3-benzylthiourea
737135
≥98%

Chemical Identifiers

13431-41-9
181.257
ZTRUHAVBRPABTK-UHFFFAOYSA-N
737135
C1=CC=C(C=C1)CNC(=S)NN
C8H11N3S
MFCD00025145
4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea
1-amino-3-benzylthiourea
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents