missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione, TCI America™

$172.64 - $514.88

Chemical Identifiers

CAS 327036-89-5
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.262
MDL Number MFCD04973552
InChI Key JDSJDASOXWCHPN-UHFFFAOYSA-N
Synonym TDZD-8
PubChem CID 4124851
IUPAC Name 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
SMILES CN1C(=O)N(C(=O)S1)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44365MG View Documents TCI America
B44365MG
5mg Each for $172.64
 
B443625MG View Documents TCI America
B443625MG
25mg Each for $514.88
 
Specifications

Chemical Identifiers

327036-89-5
222.262
JDSJDASOXWCHPN-UHFFFAOYSA-N
4124851
CN1C(=O)N(C(=O)S1)CC2=CC=CC=C2
C10H10N2O2S
MFCD04973552
TDZD-8
4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione

Specifications

White
5mg
C10H10N2O2S
TDZD-8
CN1C(=O)N(C(=O)S1)CC2=CC=CC=C2
222.262
222.26
≥98.0% (HPLC)
63°C
327036-89-5
MFCD04973552
JDSJDASOXWCHPN-UHFFFAOYSA-N
4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
4124851
Crystal-Powder at 20°C
4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)